4-Amino-3-(trifluormethyl)pyridin 97%

Lieferant: Apollo Scientific
Achtung

PC9760-1G PC9760-500MG
APOSPC9760-1GEA 252 EUR
APOSPC9760-1G APOSPC9760-500MG
4-Amino-3-(trifluormethyl)pyridin 97%
4-Amino-3-(trifluoromethyl)pyridine
Formel: C₆H₅F₃N₂
Molecular Weight: 162.11 g/mol
Schmelzpunkt: 102…103 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02183554
CAS-Nummer: 387824-61-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR