1-[4-(Trifluoromethyl)pyrimidin-2-yl]piperidine-4-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

PC9763-250MG
APOSPC9763-250MGEA 65.4 EUR
APOSPC9763-250MG
1-[4-(Trifluoromethyl)pyrimidin-2-yl]piperidine-4-carboxylic acid 97%
1-[4-(Trifluoromethyl)pyrimidin-2-yl]piperidine-4-carboxylic acid
Formel: C₁₁H₁₂F₃N₃O₂
Schmelzpunkt: 140…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02091615
CAS-Nummer: 465514-39-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR