2,4-Difluorobenzylmagnesium bromide 0,25 M in Diethylether

Lieferant: Apollo Scientific
Gefahr

PC9771-100ML
APOSPC9771-100MLEA 654 EUR
APOSPC9771-100ML
2,4-Difluorobenzylmagnesium bromide 0,25 M in Diethylether
2,4-Difluorobenzylmagnesium bromide
Formel: C₇H₅BrF₂Mg
Lagertemperatur: Raumtemperatur
MDL: MFCD07698746
CAS-Nummer: 546122-71-8
UN: 2924
ADR: 3+8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR