5-(Difluoromethoxy)-2-mercaptobenzimidazole 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Difluoromethoxy)-2-benzimidazolethiol, 5-Difluoromethoxy-2-mercaptobenzimidazole, 5-(Difluoromethoxy)-1H-benzo[d]imidazole-2-thiol, 5-(Difluoromethoxy)-2-mercapto-1H-benzimidazole

PC9818-25G
APOSPC9818-25GEA 21.9 EUR
APOSPC9818-25G
5-(Difluoromethoxy)-2-mercaptobenzimidazole 99%
5-(Difluoromethoxy)-2-mercaptobenzimidazole
Formel: C₈H₆F₂N₂OS
Molecular Weight: 216.21 g/mol
Schmelzpunkt: 239…243 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00467504
CAS-Nummer: 97963-62-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR