2-Cyan-6-fluoranilin 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC9826-1G
PC9826-25G
PC9826-5G
APOSPC9826-1GEA
21.9
EUR
APOSPC9826-1G
APOSPC9826-25G
APOSPC9826-5G
2-Cyan-6-fluoranilin 97%
2-Cyan-6-fluoranilin
Formel:
C₇H₅FN₂ Schmelzpunkt: 79…80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368774 CAS-Nummer: 115661-37-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...