4-(Trifluoromethylthio)phenyl isocyanate 97%

Lieferant: Apollo Scientific
Gefahr

PC9864-250MG
APOSPC9864-250MGEA 36.4 EUR
APOSPC9864-250MG
4-(Trifluoromethylthio)phenyl isocyanate 97%
4-(Trifluoromethylthio)phenyl isocyanate

  • May form small amount of sediment.

Formel: C₈H₄F₃NOS
Molecular Weight: 219,19 g/mol
Siedepunkt: 92…94 °C/15mmHg
Dichte: 1,385 g/cm³ (25 °C)
Flammpunkt: 80 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00036025
CAS-Nummer: 24032-84-6
UN: 2206
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR