1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12-Pentacosafluor-14-iodtetradecan 90%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Iodo-1H,1H,2H,2H-perfluorotetradecane

PC9868-1G
APOSPC9868-1GEA 161 EUR
APOSPC9868-1G
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12-Pentacosafluor-14-iodtetradecan 90%
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12-Pentacosafluor-14-iodtetradecan
Formel: C₁₄H₄F₂₅I
Molecular Weight: 774,05 g/mol
Schmelzpunkt: 110…113 °C
Lagertemperatur: Kühlschrank
CAS-Nummer: 30046-31-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR