1H-Tridecafluoro-3,3-dimethylhex-1-yne
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,3-Bis(trifluoromethyl)-4,4,5,5,6,6,6-heptafluorohex-1-yne, 3,3-Bis(trifluoromethyl)-3-(perfluoroprop-1-yl)prop-1-yne
PC9873-1G
APOSPC9873-1GEA
55.3
EUR
APOSPC9873-1G
1H-Tridecafluoro-3,3-dimethylhex-1-yne
1H-Tridecafluoro-3,3-dimethylhex-1-yne
Formel:
C₈HF₁₃ Molecular Weight: 344,073 g/mol Siedepunkt: 89 °C Dichte: 1,56 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00155807 CAS-Nummer: 261503-44-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...