4,5-Difluorobenzene-1,2-diol 97%
Lieferant: Apollo Scientific
Synonyme:
4,5-Difluorocatechol, 1,2-Difluoro-4,5-dihydroxybenzene
PC9889-250MG
APOSPC9889-250MGEA
174
EUR
APOSPC9889-250MG
4,5-Difluorobenzene-1,2-diol 97%
4,5-Difluorobenzene-1,2-diol
Formel:
C₆H₄F₂O₂ Molecular Weight: 146,092 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08458183 CAS-Nummer: 147300-08-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...