2'-Fluor-6'-hydroxyacetophenon

Lieferant: Apollo Scientific
Achtung

PC9896-5G PC9896-1G PC9896-10G
APOSPC9896-5GEA 0 EUR
APOSPC9896-5G APOSPC9896-1G APOSPC9896-10G
2'-Fluor-6'-hydroxyacetophenon
2'-Fluor-6'-hydroxyacetophenon
Formel: C₈H₇FO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01090995
CAS-Nummer: 93339-98-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR