3-Chloro-6-(trifluoromethyl)benzo[b]thiophene-2-carboxylic acid

Lieferant: Apollo Scientific

PC99009-1G
APOSPC99009-1GEA 0 EUR
APOSPC99009-1G
3-Chloro-6-(trifluoromethyl)benzo[b]thiophene-2-carboxylic acid
3-Chloro-6-(trifluoromethyl)benzo[b]thiophene-2-carboxylic acid
Formel: C₁₀H₄ClF₃O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD08444955
CAS-Nummer: 923849-73-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR