5-Bromo-1,2-difluoro-3-(methoxymethyl)benzene 95%

Lieferant: Apollo Scientific

PC99018-1G
APOSPC99018-1GEA 836 EUR
APOSPC99018-1G
5-Bromo-1,2-difluoro-3-(methoxymethyl)benzene 95%
5-Bromo-1,2-difluoro-3-(methoxymethyl)benzene
Formel: C₈H₇BrF₂O
Molecular Weight: 237,041 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD32707295
CAS-Nummer: 2484889-23-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR