2-Brom-6-fluoranilin 99%

Lieferant: Apollo Scientific
Achtung

PC9902-100G PC9902-25G PC9902-5G PC9902-1KG
APOSPC9902-100GEA 132 EUR
APOSPC9902-100G APOSPC9902-25G APOSPC9902-5G APOSPC9902-1KG
2-Brom-6-fluoranilin 99%
2-Brom-6-fluoranilin
Formel: C₆H₅BrFN
Molecular Weight: 190.01 g/mol
Dichte: 1.674 g/cm³ (25 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD01310982
CAS-Nummer: 65896-11-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR