1-Chloro-2-fluoro-3-(methoxymethyl)benzene 95%

Lieferant: Apollo Scientific

PC99024-5G
APOSPC99024-5GEA 266 EUR
APOSPC99024-5G
1-Chloro-2-fluoro-3-(methoxymethyl)benzene 95%
1-Chloro-2-fluoro-3-(methoxymethyl)benzene
Formel: C₈H₈ClFO
Molecular Weight: 174.6 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD32707293
CAS-Nummer: 2484889-18-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR