2-Methyl-4-(trifluoromethoxy)acetanilide

Lieferant: Apollo Scientific
Achtung

PC9904-1G PC9904-5G
APOSPC9904-1GEA 140 EUR
APOSPC9904-1G APOSPC9904-5G
2-Methyl-4-(trifluoromethoxy)acetanilide
2-Methyl-4-(trifluoromethoxy)acetanilide
Formel: C₁₀H₁₀F₃NO₂
Molecular Weight: 233.187 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368678
CAS-Nummer: 886762-79-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR