4-Bromo-5-fluoro-2-(trifluoromethyl)aniline

Lieferant: Apollo Scientific

PC99131-1G
APOSPC99131-1GEA 799 EUR
APOSPC99131-1G
4-Bromo-5-fluoro-2-(trifluoromethyl)aniline
4-Bromo-5-fluoro-2-(trifluoromethyl)aniline
Formel: C₇H₄BrF₄N
MDL: MFCD28740801
CAS-Nummer: 1805503-76-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR