4-Bromo-2-fluoro-3-(trifluoromethyl)benzonitrile 95%

Lieferant: Apollo Scientific
Gefahr

PC99135-1G
APOSPC99135-1GEA 522 EUR
APOSPC99135-1G
4-Bromo-2-fluoro-3-(trifluoromethyl)benzonitrile 95%
4-Bromo-2-fluoro-3-(trifluoromethyl)benzonitrile
Formel: C₈H₂BrF₄N
Molecular Weight: 268,01 g/mol
MDL: MFCD33404170
CAS-Nummer: 2090463-13-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR