6-Fluoro-4-cyanoquinoline N-oxide 95%
Lieferant: Apollo Scientific
Achtung
|
PC99144-250MG
PC99144-1G
APOSPC99144-250MGEA
227
EUR
APOSPC99144-250MG
APOSPC99144-1G
6-Fluoro-4-cyanoquinoline N-oxide 95%
6-Fluoro-4-cyanoquinoline N-oxide
Formel:
C₁₀H₅FN₂O |
MDL:
MFCD33404154 CAS-Nummer: 152562-07-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...