6-Fluoro-4-cyanoquinoline N-oxide 95%

Lieferant: Apollo Scientific
Achtung

PC99144-250MG PC99144-1G
APOSPC99144-250MGEA 227 EUR
APOSPC99144-250MG APOSPC99144-1G
6-Fluoro-4-cyanoquinoline N-oxide 95%
6-Fluoro-4-cyanoquinoline N-oxide
Formel: C₁₀H₅FN₂O
MDL: MFCD33404154
CAS-Nummer: 152562-07-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR