2,6-Difluor-4-iodanisol

Lieferant: Apollo Scientific
Achtung

PC9917-1G
APOSPC9917-1GEA 33.5 EUR
APOSPC9917-1G
2,6-Difluor-4-iodanisol
2,6-Difluor-4-iodanisol
Formel: C₇H₅F₂IO
Lagertemperatur: Raumtemperatur
MDL: MFCD07368787
CAS-Nummer: 886762-68-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR