4-Chloro-5-fluoronicotinamide 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-5-fluoropyridine-3-carboxamide

PC99183-1G
APOSPC99183-1GEA 777 EUR
APOSPC99183-1G
4-Chloro-5-fluoronicotinamide 97%
4-Chloro-5-fluoronicotinamide
Formel: C₆H₄ClFN₂O
Molecular Weight: 174.56 g/mol
MDL: MFCD28012875
CAS-Nummer: 1706432-64-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR