3-Fluor-2-hydroxypyridin

Lieferant: Apollo Scientific
Achtung

PC9928-1G PC9928-5G PC9928-25G
APOSPC9928-1GEA 21.9 EUR
APOSPC9928-1G APOSPC9928-5G APOSPC9928-25G
3-Fluor-2-hydroxypyridin
3-Fluor-2-hydroxypyridin
Formel: C₅H₄FNO
Molecular Weight: 113.09 g/mol
Schmelzpunkt: 157…161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03095042
CAS-Nummer: 1547-29-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR