3-Fluor-2-hydroxypyridin
Lieferant: Apollo Scientific
|
Achtung
|
PC9928-1G
PC9928-5G
PC9928-25G
APOSPC9928-1GEA
21.9
EUR
APOSPC9928-1G
APOSPC9928-5G
APOSPC9928-25G
3-Fluor-2-hydroxypyridin
3-Fluor-2-hydroxypyridin
Formel:
C₅H₄FNO Molecular Weight: 113.09 g/mol Schmelzpunkt: 157…161 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095042 CAS-Nummer: 1547-29-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...