1-[4-Nitro-3-(trifluoromethyl)phenyl]piperazine

Lieferant: Apollo Scientific

PC9935-1G
APOSPC9935-1GEA 150 EUR
APOSPC9935-1G
1-[4-Nitro-3-(trifluoromethyl)phenyl]piperazine
1-[4-Nitro-3-(trifluoromethyl)phenyl]piperazine
Formel: C₁₁H₁₂F₃N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04973339
CAS-Nummer: 153204-82-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR