2-Bromo-1,3,3,3-tetrafluoroprop-1-ene 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-2-fluoro-1-(trifluoromethyl)ethene

PC9937-1G
APOSPC9937-1GEA 328 EUR
APOSPC9937-1G
2-Bromo-1,3,3,3-tetrafluoroprop-1-ene 95%
2-Bromo-1,3,3,3-tetrafluoroprop-1-ene
Formel: C₃HBrF₄
Molecular Weight: 192.938 g/mol
Siedepunkt: 29…32 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03095424
CAS-Nummer: 396716-43-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR