2,6-Difluor-4-hydroxybenzaldehyd 98%

Lieferant: Apollo Scientific
Achtung

PC9940-5G
APOSPC9940-5GEA 177 EUR
APOSPC9940-5G
2,6-Difluor-4-hydroxybenzaldehyd 98%
2,6-Difluoro-4-hydroxybenzaldehyde
Formel: C₇H₄F₂O₂
Molecular Weight: 158.1 g/mol
Schmelzpunkt: 179…182 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07772042
CAS-Nummer: 532967-21-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR