2-(Trifluoroacetyl)thiazole monohydrate

Lieferant: Apollo Scientific
Achtung

PC99407-250MG PC99407-1G PC99407-5G
APOSPC99407-250MGEA 178 EUR
APOSPC99407-250MG APOSPC99407-1G APOSPC99407-5G
2-(Trifluoroacetyl)thiazole monohydrate
2-(Trifluoroacetyl)thiazole monohydrate
Formel: C₅H₂F₃NOS·H₂O
Molecular Weight: 199.15 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30742687
CAS-Nummer: 2097800-30-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR