2-(Trifluoroacetyl)thiazole monohydrate
Lieferant: Apollo Scientific
|
Achtung
|
PC99407-250MG
PC99407-1G
PC99407-5G
APOSPC99407-250MGEA
178
EUR
APOSPC99407-250MG
APOSPC99407-1G
APOSPC99407-5G
2-(Trifluoroacetyl)thiazole monohydrate
2-(Trifluoroacetyl)thiazole monohydrate
Formel:
C₅H₂F₃NOS·H₂O Molecular Weight: 199.15 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD30742687 CAS-Nummer: 2097800-30-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...