2,5-Difluorobenzhydrazide

Lieferant: Apollo Scientific
Achtung

PC9952-1G PC9952-5G
APOSPC9952-1GEA 105 EUR
APOSPC9952-1G APOSPC9952-5G
2,5-Difluorobenzhydrazide
2,5-Difluorobenzhydrazide
Formel: C₇H₆F₂N₂O
Molecular Weight: 172.14 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD02183537
CAS-Nummer: 265644-03-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR