2-Chlor-4-fluorphenylhydrazin

Lieferant: Apollo Scientific
Achtung

PC9959-5G
APOSPC9959-5GEA 517 EUR
APOSPC9959-5G
2-Chlor-4-fluorphenylhydrazin
2-Chlor-4-fluorphenylhydrazin
Formel: C₆H₆ClFN₂
Lagertemperatur: Kühlschrank
MDL: MFCD03094567
CAS-Nummer: 119452-65-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR