5-(2-Fluorphenyl)isoxazol

Lieferant: Apollo Scientific
Achtung

PC9960-250MG
APOSPC9960-250MGEA 43.5 EUR
APOSPC9960-250MG
5-(2-Fluorphenyl)isoxazol
5-(2-Fluorophenyl)isoxazole
Formel: C₉H₆FNO
Molecular Weight: 163,15 g/mol
Siedepunkt: 68…70 °C/0,3mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD02183542
CAS-Nummer: 138716-36-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR