8-(Trifluoromethyl)quinoline-4-thiol

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Mercapto-8-(trifluoromethyl)quinoline

PC9963-1G
APOSPC9963-1GEA 244 EUR
APOSPC9963-1G
8-(Trifluoromethyl)quinoline-4-thiol
8-(Trifluoromethyl)quinoline-4-thiol
Formel: C₁₀H₆F₃NS
Molecular Weight: 229,222 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00665836
CAS-Nummer: 550346-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR