5-(2,4-Difluorphenyl)isoxazol

Lieferant: Apollo Scientific
Achtung

PC9964-250MG
APOSPC9964-250MGEA 58 EUR
APOSPC9964-250MG
5-(2,4-Difluorphenyl)isoxazol
5-(2,4-Difluorophenyl)isoxazole
Formel: C₉H₅F₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD02183538
CAS-Nummer: 138716-46-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR