3-(4-Fluorophenylthio)propanoic acid, tech
Lieferant: Apollo Scientific
|
Gefahr
|
PC9991-1G
APOSPC9991-1GEA
26.1
EUR
APOSPC9991-1G
3-(4-Fluorophenylthio)propanoic acid, tech
3-(4-Fluorophenylthio)propanoic acid
Formel:
C₉H₉FO₂S Molecular Weight: 200,23 g/mol Schmelzpunkt: 75…78 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236613 CAS-Nummer: 19543-85-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...