Azomethin H Reag. Ph. Eur. analytisches Reagens
Lieferant: PanReac AppliChem
Synonyme:
8-Hydroxy-1-[[(2-hydroxyphenyl)methylen]amino]naphthalin-3,6-disulfonsäure, Azomethine-H, 8-Hydroxy-1-(salicylidenamino)naphthalen-3,6-disulfonsäure, 4-Hydroxy-5-[[(2-hydroxyphenyl)methylen]amino]naphthalin-2,7-disulfonsäure, 4-Hydroxy-5-(salicylidenamino)naphthalin-2,7-disulfonsäure
123581.1605
APPC123581.1605EA
115
EUR
APPC123581.1605
Azomethin H Reag. Ph. Eur. analytisches Reagens
Azomethin H
Azomethine H acts as boron reagent.
Synonyms: 4-Hydroxy-5-(2-Hydroxybenzylidenamine)-Naphtalene-2; 7-Disulphonic Acid.
Formel:
C₁₇H₁₃NO₈S₂ Molecular Weight: 423,42 g/mol |
CAS-Nummer:
32266-60-7 |
Spezifikation Testergebnisse
Identity | IR passes test |
Maximum limit of impurities (Sensitivity to B) | Passes test |
Absorbance (1%) at λ236 nm at (pH 5,1) | >1200 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...