Kaliumdihydrogenphosphat ≥99,0% (durch Acidimetrie) ACS analytisches Reagens

Lieferant: PanReac AppliChem

131509.1210 131509.1211 131509.1214
APPC131509.1210EA 37.9 EUR
APPC131509.1210 APPC131509.1211 APPC131509.1214
Kaliumdihydrogenphosphat ≥99,0% (durch Acidimetrie) ACS analytisches Reagens
Kaliumdihydrogenphosphat
Formel: KH₂PO₄
Siedepunkt: 252 °C
Schmelzpunkt: 252 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011401
CAS-Nummer: 7778-77-0
EINECS: 231-913-4

Bestellen

Spezifikation Testergebnisse

Minimum Assay (Acidim.) 99.0%
pH of 5% Solution 4.2 - 4.5
Insoluble Matter in H₂O Max. 0.01%
Loss on Drying at 105 °C Max. 0.2%
Chloride (Cl) Max. 0.0005%
Nitrogen Compounds (as N) Max. 0.001%
Sulphate (SO₄) Max. 0.003%
As Max. 0.0002%
Heavy Metals (ICP-OES) Max. 0.001%
Ag (by ICP) 5 mg/kg (ppm)
Al (by ICP) 5 mg/kg (ppm)
Au (by ICP) 5 mg/kg (ppm)
B (by ICP) 5 mg/kg (ppm)
Ba (by ICP) 5 mg/kg (ppm)
Be (by ICP) 5 mg/kg (ppm)
Bi (by ICP) 5 mg/kg (ppm)
Ca (by ICP) 50 mg/kg (ppm)
Cd (by ICP) 5 mg/kg (ppm)
Co (by ICP) 5 mg/kg (ppm)
Cr (by ICP) 5 mg/kg (ppm)
Cu (by ICP) 5 mg/kg (ppm)
Fe (by ICP) 10 mg/kg (ppm)
Ga (by ICP) 5 mg/kg (ppm)
Ge (by ICP) 5 mg/kg (ppm)
In (by ICP) 5 mg/kg (ppm)
Mg (by ICP) 5 mg/kg (ppm)
Mn (by ICP) 5 mg/kg (ppm)
Mo (by ICP) 5 mg/kg (ppm)
Na (by ICP) 50 mg/kg (ppm)
Ni (by ICP) 5 mg/kg (ppm)
Pb (by ICP) 5 mg/kg (ppm)
Sb (by ICP) 5 mg/kg (ppm)
Se (by ICP) 5 mg/kg (ppm)
Si (by ICP) 5 mg/kg (ppm)
Sr (by ICP) 5 mg/kg (ppm)
Ti (by ICP) 5 mg/kg (ppm)
Tl (by ICP) 5 mg/kg (ppm)
V (by ICP) 5 mg/kg (ppm)
Zn (by ICP) 5 mg/kg (ppm)
Zr (by ICP) 5 mg/kg (ppm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR