Kaliumdihydrogenphosphat ≥99,0% (durch Acidimetrie) ACS analytisches Reagens
Lieferant: PanReac AppliChem
131509.1210
131509.1211
131509.1214
APPC131509.1210EA
37.9
EUR
APPC131509.1210
APPC131509.1211
APPC131509.1214
Kaliumdihydrogenphosphat ≥99,0% (durch Acidimetrie) ACS analytisches Reagens
Kaliumdihydrogenphosphat
Formel:
KH₂PO₄ Siedepunkt: 252 °C Schmelzpunkt: 252 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011401 CAS-Nummer: 7778-77-0 EINECS: 231-913-4 |
Spezifikation Testergebnisse
Minimum Assay (Acidim.) | 99.0% |
pH of 5% Solution | 4.2 - 4.5 |
Insoluble Matter in H₂O | Max. 0.01% |
Loss on Drying at 105 °C | Max. 0.2% |
Chloride (Cl) | Max. 0.0005% |
Nitrogen Compounds (as N) | Max. 0.001% |
Sulphate (SO₄) | Max. 0.003% |
As | Max. 0.0002% |
Heavy Metals (ICP-OES) | Max. 0.001% |
Ag (by ICP) | 5 mg/kg (ppm) |
Al (by ICP) | 5 mg/kg (ppm) |
Au (by ICP) | 5 mg/kg (ppm) |
B (by ICP) | 5 mg/kg (ppm) |
Ba (by ICP) | 5 mg/kg (ppm) |
Be (by ICP) | 5 mg/kg (ppm) |
Bi (by ICP) | 5 mg/kg (ppm) |
Ca (by ICP) | 50 mg/kg (ppm) |
Cd (by ICP) | 5 mg/kg (ppm) |
Co (by ICP) | 5 mg/kg (ppm) |
Cr (by ICP) | 5 mg/kg (ppm) |
Cu (by ICP) | 5 mg/kg (ppm) |
Fe (by ICP) | 10 mg/kg (ppm) |
Ga (by ICP) | 5 mg/kg (ppm) |
Ge (by ICP) | 5 mg/kg (ppm) |
In (by ICP) | 5 mg/kg (ppm) |
Mg (by ICP) | 5 mg/kg (ppm) |
Mn (by ICP) | 5 mg/kg (ppm) |
Mo (by ICP) | 5 mg/kg (ppm) |
Na (by ICP) | 50 mg/kg (ppm) |
Ni (by ICP) | 5 mg/kg (ppm) |
Pb (by ICP) | 5 mg/kg (ppm) |
Sb (by ICP) | 5 mg/kg (ppm) |
Se (by ICP) | 5 mg/kg (ppm) |
Si (by ICP) | 5 mg/kg (ppm) |
Sr (by ICP) | 5 mg/kg (ppm) |
Ti (by ICP) | 5 mg/kg (ppm) |
Tl (by ICP) | 5 mg/kg (ppm) |
V (by ICP) | 5 mg/kg (ppm) |
Zn (by ICP) | 5 mg/kg (ppm) |
Zr (by ICP) | 5 mg/kg (ppm) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...