Aceton ≥99,5% (durch GC), Flüssigkeit USP, Ph. Eur., BP, rein
Lieferant: PanReac AppliChem
|
Gefahr
|
Synonyme:
Propanon, DMK, 2-Propanon, Dimethylketon
141007.1611
141007.1214
141007.0716
141007.0816
141007.1211
141007.1212
APPC141007.1611EA
32.1
EUR
APPC141007.1611
APPC141007.1214
APPC141007.0716
APPC141007.0816
APPC141007.1211
APPC141007.1212
Aceton ≥99,5% (durch GC), Flüssigkeit USP, Ph. Eur., BP, rein
Aceton
Synonyms: 2-Propanone, ß-Ketopropane, Dimethylketone, Pyroacetic Ether.
Formel:
CH₃COCH₃ Siedepunkt: 56,5 °C Dichte: 0,789 Kg/l |
MDL:
MFCD00008765 CAS-Nummer: 67-64-1 UN: 1090 ADR: ,Ⅱ |
|
Spezifikation Testergebnisse
Solubility | Water |
Refractive index | 20/D 1,3591 |
Appearance | Liquid |
Min. assay (G.C.) | 99,5% |
Identity according to pharmacopoeias | Passes test |
Density 25/25 | ≤0,789 |
Density 20/20 | 0,790 - 0,793 |
Appearance of solution | Passes test |
Acidity | Passes test |
Alkalinity | Passes test |
Insoluble matter in H₂O | Passes test |
Non-volatile matter | 0,004% |
Reducing substance to KMnO₄ | Passes test |
Residual solvents (Ph. Eur./USP) | Passes test |
Methanol | 0,05% |
2-Propanol | 0,05% |
Benzene | 0,0002% |
Other impurities | 0,05% |
Ethanol (G.C.) | 500 ppm |
Water (H₂O) | 0,3% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...