Calciumacetat 99,0-100,5% USP
Lieferant: PanReac AppliChem
Synonyme:
Essigsäure Calciumsalz, Calciumdi(acetat)
141211.1211
APPC141211.1211EA
84.2
EUR
APPC141211.1211
Calciumacetat 99,0-100,5% USP
Calciumacetat
Formel:
(H₃CCOO)₂Ca Molecular Weight: 158,17 g/mol |
MDL:
MFCD00150019 CAS-Nummer: 62-54-4 EINECS: 200-540-9 |
Spezifikation Testergebnisse
Assay (Compl.) (calc. a.a.s) | 99,0-100,5% |
Presentation | White Small Crystals |
Solubility | Soluble in water 300 g/l at 20 °C, Hygroscopic |
Termal Stability | Thermal decomposition above 160 °C |
pH (0,2 M sol.) | 7,6 |
pH of 5% solution | 6,3-9,6 |
Insoluble matter in H2O | 0,1% Max. |
Easily oxidable substances | Passes Test |
Chloride (Cl) | 0,05% Max. |
Phosphate(PO4) | 0,005% Max. |
Sulphate (SO4) | 0,06% Max. |
Residual solvents (Ph. Eur/USP) | Passes Test |
Fluoride (F) | 0,005% Max. |
Nitrate (NO3) | Passes Test |
Water (H2O) | 7,0% Max. |
Heavy metals (as Pb) | 0,002% Max. |
Aluminium (Al) | 0,001% Max. |
Arsenic (As) | 0,0002% Max. |
Barium (Ba) | 0,005% Max. |
Copper (Cu) | 0,001% Max. |
Iron (Fe) | 0,005% Max. |
Magnesium (Mg) | 0,05% Max. |
Nickel (Ni) | 0,001% Max. |
Lead (Pb) | 0,001% Max. |
Strontium (Sr) | 0,05% Max. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...