Eisen(III)chlorid Hexahydrat 97-102% (durch Iodometrie), rein

Lieferant: PanReac AppliChem

Synonyme: Eisentrichlorid Hexahydrat

141358.1214
APPC141358.1214EA 0 EUR
APPC141358.1214
Eisen(III)chlorid Hexahydrat 97-102% (durch Iodometrie), rein
Eisen(III)chlorid Hexahydrat
Formel: FeCl₃·6H₂O
Molecular Weight: 270.3 g/mol
Schmelzpunkt: 37
MDL: MFCD00149712
CAS-Nummer: 10025-77-1
UN: 3260
ADR: 8 III
Merck Index: 13,04048
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=808c75376e55f29296d58eb1e5a1e184039ce733&vid=6d820873011bee3ff401441544f7f85599e40626&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (Iodometr.) 97 - 102%
Acidity Passes Test
P Compounds (as PO₄) Max. 0.05%
Sulphate Max. 0.05%
Ca Max. 0.1%
Cu Max. 0.015%
As Max. 0.001%
PB Max. 0.005%
K Max. 0.05%
NA Max. 0.1%
Zn Max. 0.01%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR