Xylol (Isomerengemisch) ≥98%, Flüssigkeit

Lieferant: PanReac AppliChem

Synonyme: Dimethylbenzol

141769.0314 141769.1612 141769.2711 141769.3514
APPC141769.0314EA 141 EUR
APPC141769.0314 APPC141769.1612 APPC141769.2711 APPC141769.3514
Xylol (Isomerengemisch) ≥98%, Flüssigkeit
Xylol (Isomerengemisch)

Isomer mixture, mainly m-Xylene ca. 70%, Ethylbenzene 20% and variable amounts of o- and p-Xilene.


Synonyms: Dimethylbenzene, xilol.

Formel: C₆H₄(CH₃)₂
Molecular Weight: 106.17 g/mol
Siedepunkt: 137 – 144 °C
Flammpunkt: 29 °C
MDL: MFCD00077264
CAS-Nummer: 1330-20-7
UN: 1307
ADR: 3 III
Merck Index: 13,10136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearence Clear colorless liquid
Solubility Soluble in water 0,2 g/l at 20 °C
Vapor pressure 20 °C
Assay 98%
Refractive index n20/D 1,497
Ignition temperature 465 °C
Vapour pressure (20 °C) 8 hPa
Dielectric constant (25 °C) 2,4
Evaporation number (DIN 53170) 13,5
Saturation concentration (20 °C) 35 g/m³
Explosion limit 1,7 − 7,6% (V)
Identity (IR) Passes test
Density at 20/4 0,862 – 0,864
Acidity 0,0003 meq/g
Alkalinity 0,00025 meq/g
Non-volatile matter 0,01%
Thiophene (C₄H₄S) 0,0005%
Water (H₂O) 0,05%
Copper (Cu) 0,00002%
Iron (Fe) 0,00005%
Nickel (Ni) 0,00002%
Lead (Pb) 0,00002%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR