Tetradecyltrimethylammoniumbromid BP, Ph.Eur., rein, Pharmaqualität
Lieferant: PanReac AppliChem
|
Gefahr
|
Synonyme:
Myristyltrimethylammoniumbromid, Tetradoniumbromid, TTAB, Dynamin Inhibitor II
142542.1209
APPC142542.1209EA
78.5
EUR
APPC142542.1209
Tetradecyltrimethylammoniumbromid BP, Ph.Eur., rein, Pharmaqualität
Tetradecyltrimethylammoniumbromid
Formel:
C₁₇H₃₈BrN Molecular Weight: 336,394 g/mol |
MDL:
MFCD00011770 CAS-Nummer: 8044-71-1 UN: 1759 ADR: 8,II |
Spezifikation Testergebnisse
Solubility | Water 500 g/L at 20 °C |
Appearance of solution | Passes test |
Amines and amine salts | Passes test |
Acidity or alkalinity | Passes test |
Insoluble matter in H₂O | Passes test |
Loss on drying at 105 °C | 2,0% |
Residue on ignition (as SO₄) | 0,5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...