Ölsäure

Lieferant: PanReac AppliChem
Achtung

Synonyme: cis-9-Octadecensäure

142659.0716 142659.1611
APPC142659.0716EA 0 EUR
APPC142659.0716 APPC142659.1611
Ölsäure
Ölsäure

Synonyms: (Z)-9-Octadecenoic acid.

Formel: C₁₈H₃₄O₂
Molecular Weight: 282.47 g/mol
Siedepunkt: 286 °C
Schmelzpunkt: 4 °C
Dichte: 0.890 kg/l
Flammpunkt: 180
MDL: MFCD00064242
CAS-Nummer: 112-80-1
Merck Index: 13,06898
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5aecc5dce4d0318386a45d7adb7c5822631010a&vid=892a2df4ecab3679b06983b84cd27ad334ef961f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (G.C.) 65,0 - 88,0%
Appearance Passes test
Residue on ignition (as SO₄) 0,1%
Acidity value 195 - 204
Iodine value 89 - 105
Peroxide value 10,0
Residual solvents (Ph. Eur.) Passes test
Myristic acid 5,0%
Palmitic acid 16,0%
Palmitoleic acid 8,0%
Stearic acid 6,0%
Linoleic acid 18,0%
Linolenic acid 4,0%
Fatty acids with chain length greater than C18 4,0%
Margaric acid 4,0%
Identity IR passes test
Cadmium (Cd) - Class 1 impurities (ICH Q3D) 0,5 ppm
Lead (Pb) - Class 1 impurities (ICH Q3D) 2 ppm
Arsenic (As) - Class 1 impurities (ICH Q3D) 1,5 ppm
Mercury (Hg) - Class 1 impurities (ICH Q3D) 1,5 ppm
Cobalt (Co) - Class 2A impurities (ICH Q3D) 5 ppm
Vanadium (V) - Class 2A impurities (ICH Q3D) 10 ppm
Nickel (Ni) - Class 2A impurities (ICH Q3D) 20 ppm
Thallium (Tl) - Class 2B impurities (ICH Q3D) 5 ppm
Gold (Au) - Class 2B impurities (ICH Q3D) 10 ppm
Palladium (Pd) - Class 2B impurities (ICH Q3D) 10 ppm
Iridium (Ir) - Class 2B impurities (ICH Q3D) 10 ppm
Osmium (Os) - Class 2B impurities (ICH Q3D) 10 ppm
Rhodium (Rh) - Class 2B impurities (ICH Q3D) 10 ppm
Ruthenium (Ru) - Class 2B impurities (ICH Q3D) 10 ppm
Selenium (Se) - Class 2B impurities (ICH Q3D) 15 ppm
Silver (Ag) - Class 2B impurities (ICH Q3D) 15 ppm
Platinum (Pt) - Class 2B impurities (ICH Q3D) 10 ppm
Lithium (Li) - Class 3 impurities (ICH Q3D) 55 ppm
Antimony (Sb) - Class 3 impurities (ICH Q3D) 120 ppm
Barium (Ba) - Class 3 impurities (ICH Q3D) 140 ppm
Molybdenum (Mo) - Class 3 impurities (ICH Q3D) 25 ppm
Copper (Cu) - Class 3 impurities (ICH Q3D) 250 ppm
Tin (Sn) - Class 3 impurities (ICH Q3D) 600 ppm
Chromium (Cr) - Class 3 impurities (ICH Q3D) 25 ppm

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR