Salzsäure 0.1 M (0.1 N) Volumetric
Lieferant: PanReac AppliChem
Synonyme:
Hydrochloric acid solution, Muriatic acid
181023.1212
181023.0715
181023.1211
181023.1214
181023.1315
APPC181023.1212EA
53.9
EUR
APPC181023.1212
APPC181023.0715
APPC181023.1211
APPC181023.1214
APPC181023.1315
Salzsäure 0.1 M (0.1 N) Volumetric
Salzsäure
- Miscible with water
Standards: Traceable to NIST.
Formel:
HCl Molecular Weight: 36.46 g/mol |
MDL:
MFCD00011324 CAS-Nummer: 7647-01-0 UN: 1789 ADR: 8 III |
Spezifikation Testergebnisse
Molarity | 0,1 M |
Appearence | Clear colourless liquid |
pH | 1,2 |
Titer at 20 °C | 0,999 – 1,001 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...