Silbernitrat 0.1 M (0.1 N) Reag. USP volumetrische Lösung

Lieferant: PanReac AppliChem

186983.1211
APPC186983.1211EA 141 EUR
APPC186983.1211
Silbernitrat 0.1 M (0.1 N) Reag. USP volumetrische Lösung
Silbernitrat

Standardised against sodium chloride using eosine indicator.

Formel: AgNO₃
MDL: MFCD00003414
CAS-Nummer: 7761-88-8
EINECS: 231-853-9
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Presentation Clear Colourless Liquid
Solubility Miscible with water (D 20/4) 1,012
pH 4 -5
Titer at 20 °C 0,999 - 1,001
Traceability NIST


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR