Wasser, technische Qualität
Lieferant: PanReac AppliChem
211074.0715
211074.0716
211074.1214
APPC211074.0715EA
44.4
EUR
APPC211074.0715
APPC211074.0716
APPC211074.1214
Wasser, technische Qualität
Wasser
Manufactured by reverse osmosis has purity level comparable to two times distilled water.
Formel:
H₂O Siedepunkt: 100 °C Schmelzpunkt: 0 °C Dichte: 1,00 kg/l (20 ˚C) |
MDL:
MFCD00011332 CAS-Nummer: 7732-18-5 |
Spezifikation Testergebnisse
Form | Liquid |
Sterility | Not tested |
Conductivity | Max. 5,0 μS/cm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...