Bouin liquor
Lieferant: PanReac AppliChem
254102.1611
APPC254102.1611EA
75.5
EUR
APPC254102.1611
Bouin liquor
Färbemittel
Mikroskopie- und Färbereagenzien
Fixative for preserving soft and delicate structures, used as a mordant in various trichrome procedures.
Composition:
Picric Acid moistened with ~33% H₂O: 1,125 g
Acetic Acid glacial: 5 ml
Formaldehyde 35 to 40%: 25 ml
Water: 77 ml
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...