Buffer solution, pH 12,454
Lieferant: PanReac AppliChem
277089.1209
APPC277089.1209EA
24.1
EUR
APPC277089.1209
Buffer solution, pH 12,454
Puffer
pH-Puffer
These buffer solutions are useful for the calibration of high precision laboratory pH meters.
- pH (30 °C): 12,289 ±0,01
- pH (25 °C): 12,454 ±0,01
- pH (20 °C): 12,627 ±0,01
- pH (10 °C): 12,810 ±0,01
Standards: Standardised, according to DIN19266.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...