Buffer solution, pH 6,865
Lieferant: PanReac AppliChem
277124.1211
APPC277124.1211EA
49.6
EUR
APPC277124.1211
Buffer solution, pH 6,865
Puffer
pH-Puffer
These buffer solutions are useful for the calibration of high precision laboratory pH meters.
- pH (30 °C): 6,853 ± 0,01
- pH (25 °C): 6,865 ± 0,01
- pH (20 °C): 6,881 ± 0,01
- pH (10 °C): 6,900 ± 0,01
Standards: Standardised, according to DIN19266.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...