Folsäure
Lieferant: PanReac AppliChem
Synonyme:
Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9
A7092.0100
A7092.0025
APPCA7092.0100EA
257
EUR
APPCA7092.0100
A7092.0025
Folsäure
Folsäure
Formel:
C₁₉H₁₉N₇O₆ Molecular Weight: 441.41 g/mol |
MDL:
MFCD00079305 CAS-Nummer: 59-30-3 EINECS: 200-419-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...